Assay Details
| Gene Symbol: Lnx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496660.2 | XP_003496708.1 | 8 - 9 | 1980 | 85 | |
| XM_007652163.1 | XP_007650353.1 | 9 - 10 | 2164 | 85 | ||
| XM_007652170.1 | XP_007650360.1 | 8 - 9 | 1999 | 85 | ||
| XM_007652175.1 | XP_007650365.1 | 8 - 9 | 1847 | 85 | ||
More Information
Set Membership: |
|

Lnx2
Genomic Map