Assay Details
| Gene Symbol: Sec14l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509013.1 | XP_003509061.1 | 6 - 7 | 520 | 67 | |
| XM_007649650.1 | XP_007647840.1 | 6 - 7 | 520 | 67 | ||
| XM_007649651.1 | XP_007647841.1 | 6 - 7 | 520 | 67 | ||
More Information
Set Membership: |
|

Sec14l3
Genomic Map