Assay Details
| Gene Symbol: Pdpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509536.2 | XP_003509584.2 | 12 - 13 | 1402 | 97 | |
| GenBank mRNA | JP039402.1 | - | 7 - 8 | 790 | 97 | |
More Information
Set Membership: |
|

Pdpk1
Genomic Map