Assay Details
| Gene Symbol: Ckmt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498034.2 | XP_003498082.1 | 1 - 2 | 345 | 69 | |
| XM_003498035.2 | XP_003498083.1 | 2 - 3 | 287 | 69 | ||
| XM_007621602.1 | XP_007619792.1 | 2 - 3 | 339 | 69 | ||
More Information
Set Membership: |
|

Ckmt1b
Genomic Map