Assay Details
| Gene Symbol: Gyltl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497286.1 | XP_003497334.1 | 10 - 11 | 1238 | 53 | |
| XM_003497287.1 | XP_003497335.1 | 9 - 10 | 1133 | 53 | ||
| XM_007609903.1 | XP_007608093.1 | 10 - 11 | 1256 | 53 | ||
More Information
Set Membership: |
|

Gyltl1b
Genomic Map