Assay Details
| Gene Symbol: Tomm20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496800.2 | XP_003496848.1 | 4 - 5 | 502 | 74 | |
| GenBank mRNA | JI876866.1 | - | 4 - 5 | 502 | 74 | |
| JP044741.1 | - | 4 - 5 | 502 | 74 | ||
More Information
Set Membership: |
|

Tomm20
Genomic Map