Assay Details
| Gene Symbol: Timm10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512859.2 | XP_003512907.1 | 2 - 3 | 205 | 104 | |
| XM_007653262.1 | XP_007651452.1 | 2 - 3 | 205 | 104 | ||
| GenBank mRNA | JI868134.1 | - | 2 - 3 | 186 | 104 | |
| JP053350.1 | - | 2 - 3 | 186 | 104 | ||
More Information
Set Membership: |
|

Timm10b
Genomic Map