Assay Details
| Gene Symbol: Gng3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514303.2 | XP_003514351.1 | 3 - 4 | 564 | 78 | |
| XM_007654464.1 | XP_007652654.1 | 3 - 4 | 441 | 78 | ||
| GenBank mRNA | JI888003.1 | - | 2 - 3 | 246 | 79 | |
| JP054227.1 | - | 2 - 3 | 246 | 79 | ||
More Information
Set Membership: |
|

Gng3
Genomic Map