Assay Details
| Gene Symbol: Edar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503764.1 | XP_003503812.1 | 1 - 2 | 52 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Edar| Gene Symbol: Edar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503764.1 | XP_003503812.1 | 1 - 2 | 52 | 59 | |
Set Membership: |
|