Assay Details
| Gene Symbol: Spib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510893.1 | XP_003510941.1 | 5 - 6 | 508 | 78 | |
| XM_007651495.1 | XP_007649685.1 | 5 - 6 | 511 | 78 | ||
More Information
Set Membership: |
|

Spib
Genomic Map