Assay Details
| Gene Symbol: Med22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511728.2 | XP_003511776.1 | 3 - 4 | 451 | 94 | |
| GenBank mRNA | JI877558.1 | - | 1 - 2 | 170 | 94 | |
More Information
Set Membership: |
|

Med22
Genomic Map