Assay Details
| Gene Symbol: Ptp4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500644.2 | XP_003500692.1 | 3 - 4 | 858 | 124 | |
| GenBank mRNA | JP058064.1 | - | 3 - 4 | 835 | 124 | |
More Information
Set Membership: |
|

Ptp4a2
Genomic Map