Assay Details
| Gene Symbol: LOC100767915 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649568.1 | XP_007647758.1 | 1 - 2 | 285 | 81 | |
| GenBank mRNA | JP040438.1 | - | 1 - 2 | 285 | 81 | |
More Information
Set Membership: |
|

LOC100767915
Genomic Map