Assay Details
| Gene Symbol: Nfatc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501848.2 | XP_003501896.2 | 8 - 9 | 2234 | 66 | |
| XM_007642484.1 | XP_007640674.1 | 8 - 9 | 2231 | 66 | ||
More Information
Set Membership: |
|

Nfatc1
Genomic Map