Assay Details
| Gene Symbol: Skp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498487.2 | XP_003498535.1 | 3 - 4 | 194 | 101 | |
| XM_007628572.1 | XP_007626762.1 | 3 - 4 | 193 | 101 | ||
| XM_007628581.1 | XP_007626771.1 | 3 - 4 | 193 | 101 | ||
| XM_007628591.1 | XP_007626781.1 | 3 - 4 | 194 | 101 | ||
| XM_007628598.1 | XP_007626788.1 | 3 - 4 | 190 | 101 | ||
| GenBank mRNA | JI862556.1 | - | 2 - 3 | 282 | 101 | |
| JP048669.1 | - | 2 - 3 | 282 | 101 | ||
More Information
Set Membership: |
|

Skp1
Genomic Map