Assay Details
| Gene Symbol: Ppp1cc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498462.2 | XP_003498510.1 | 4 - 5 | 760 | 105 | |
| GenBank mRNA | JP052873.1 | - | 4 - 5 | 737 | 105 | |
More Information
Set Membership: |
|

Ppp1cc
Genomic Map