Assay Details
| Gene Symbol: Hoxb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640347.1 | XP_007638537.1 | 7 - 8 | 1051 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxb8| Gene Symbol: Hoxb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640347.1 | XP_007638537.1 | 7 - 8 | 1051 | 91 | |
Set Membership: |
|