Assay Details
| Gene Symbol: Mt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244576.1 | NP_001231505.1 | 1 - 2 | 31 | 67 | |
| GenBank mRNA | JI877540.1 | - | 1 - 2 | 204 | 67 | |
| JP043447.1 | - | 1 - 2 | 204 | 67 | ||
More Information
Set Membership: |
|

Mt1
Genomic Map