Assay Details
| Gene Symbol: Casp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246704.1 | NP_001233633.1 | 9 - 10 | 1280 | 55 | |
| GenBank mRNA | EU527787.1 | - | 9 - 10 | 1280 | 55 | |
More Information
Set Membership: |
|

Casp2
Genomic Map