Assay Details
| Gene Symbol: Tcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246733.1 | NP_001233662.1 | 2 - 3 | 236 | 63 | |
| GenBank mRNA | JI868660.1 | - | 2 - 3 | 322 | 63 | |
| JP051857.1 | - | 2 - 3 | 311 | 63 | ||
| M34665.1 | - | 2 - 3 | 236 | 63 | ||
More Information
Set Membership: |
|

Tcp1
Genomic Map