Assay Details
| Gene Symbol: B4galt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246777.2 | NP_001233706.1 | 3 - 4 | 744 | 73 | |
| XM_007647378.1 | XP_007645568.1 | 5 - 6 | 1020 | 73 | ||
| XM_007647379.1 | XP_007645569.1 | 5 - 6 | 944 | 73 | ||
| XM_007647380.1 | XP_007645570.1 | 5 - 6 | 938 | 73 | ||
| XM_007647381.1 | XP_007645571.1 | 5 - 6 | 880 | 73 | ||
| XM_007647382.1 | XP_007645572.1 | 4 - 5 | 851 | 73 | ||
| GenBank mRNA | AY117537.1 | - | 3 - 4 | 744 | 73 | |
| JI885640.1 | - | 3 - 4 | 729 | 73 | ||
| JP058930.1 | - | 3 - 4 | 729 | 73 | ||
More Information
Set Membership: |
|

B4galt3
Genomic Map