Assay Details
| Gene Symbol: Neu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246800.1 | NP_001233729.1 | 4 - 5 | 1014 | 86 | |
| GenBank mRNA | FJ476073.1 | - | 4 - 5 | 1014 | 86 | |
| JI895720.1 | - | 4 - 5 | 775 | 86 | ||
| JP057040.1 | - | 4 - 5 | 775 | 86 | ||
More Information
Set Membership: |
|

Neu1
Genomic Map