Assay Details
| Gene Symbol: Lrrc51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282002.1 | NP_001268931.1 | 3 - 4 | 293 | 69 | |
| NM_001282003.1 | NP_001268932.1 | 2 - 3 | 230 | 69 | ||
| GenBank mRNA | JI877748.1 | - | 1 - 2 | 138 | 69 | |
| JP062696.1 | - | 1 - 2 | 138 | 69 | ||
More Information
Set Membership: |
|

Lrrc51
Genomic Map