Assay Details
| Gene Symbol: Ahr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243990.1 | NP_001230919.1 | 5 - 6 | 712 | 64 | |
| GenBank mRNA | AB042238.1 | - | 5 - 6 | 712 | 64 | |
| JI877077.1 | - | 5 - 6 | 647 | 64 | ||
| JP061058.1 | - | 5 - 6 | 647 | 64 | ||
More Information
Set Membership: |
|

Ahr
Genomic Map