Assay Details
| Gene Symbol: Rpsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244033.1 | NP_001230962.1 | 1 | 554 | 71 | |
| GenBank mRNA | L16589.1 | - | 1 | 554 | 71 | |
| Z22749.1 | - | 1 | 554 | 71 | ||
More Information
Set Membership: |
|

Rpsa
Genomic Map