Assay Details
| Gene Symbol: Orc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246688.1 | NP_001233617.1 | 6 - 7 | 1215 | 68 | |
| XM_007650577.1 | XP_007648767.1 | 7 - 8 | 1340 | 68 | ||
| XM_007650583.1 | XP_007648773.1 | 6 - 7 | 1234 | 68 | ||
| GenBank mRNA | AF254572.1 | - | 6 - 7 | 1215 | 68 | |
| JI885369.1 | - | 6 - 7 | 1231 | 68 | ||
| JP057551.1 | - | 6 - 7 | 1231 | 68 | ||
More Information
Set Membership: |
|

Orc1
Genomic Map