Assay Details
| Gene Symbol: Htr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246728.1 | NP_001233657.1 | 1 - 2 | 1582 | 68 | |
| GenBank mRNA | X53791.1 | - | 1 - 2 | 1582 | 68 | |
More Information
Set Membership: |
|

Htr2a
Genomic Map