Assay Details
| Gene Symbol: Slc35a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246755.1 | NP_001233684.1 | 5 - 6 | 998 | 107 | |
| GenBank mRNA | JI891960.1 | - | 5 - 6 | 893 | 107 | |
| JP039706.1 | - | 6 - 7 | 894 | 107 | ||
| Y12074.1 | - | 5 - 6 | 998 | 107 | ||
More Information
Set Membership: |
|

Slc35a1
Genomic Map