Assay Details
| Gene Symbol: Hspa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246829.1 | NP_001233758.1 | 8 - 9 | 1180 | 65 | |
| GenBank mRNA | JI875975.1 | - | 8 - 9 | 1055 | 65 | |
| JP065007.1 | - | 8 - 9 | 1055 | 65 | ||
| U92313.1 | - | 8 - 9 | 1180 | 65 | ||
More Information
Set Membership: |
|

Hspa9
Genomic Map