Assay Details
| Gene Symbol: Adss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001292059.1 | NP_001278988.1 | 6 - 7 | 767 | 82 | |
| XM_007640880.1 | XP_007639070.1 | 6 - 7 | 767 | 82 | ||
| GenBank mRNA | JP049208.1 | - | 6 - 7 | 454 | 82 | |
| KF574260.1 | - | 6 - 7 | 767 | 82 | ||
More Information
Set Membership: |
|

Adss
Genomic Map