Assay Details
| Gene Symbol: Slc1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246722.1 | NP_001233651.1 | 3 - 4 | 521 | 66 | |
| XM_007644516.1 | XP_007642706.1 | 4 - 5 | 777 | 66 | ||
| GenBank mRNA | EF155647.1 | - | 3 - 4 | 521 | 66 | |
More Information
Set Membership: |
|

Slc1a3
Genomic Map