Assay Details
| Gene Symbol: Elk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246732.1 | NP_001233661.1 | 3 - 4 | 1180 | 61 | |
| GenBank mRNA | DQ384867.1 | - | 3 - 4 | 1180 | 61 | |
| JI871851.1 | - | 2 - 3 | 920 | 61 | ||
| JP045397.1 | - | 2 - 3 | 920 | 61 | ||
More Information
Set Membership: |
|

Elk3
Genomic Map