Assay Details
| Gene Symbol: Akr1b8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246751.1 | NP_001233680.1 | 9 - 10 | 945 | 79 | |
| GenBank mRNA | JI878314.1 | - | 9 - 10 | 1076 | 79 | |
| JP064643.1 | - | 9 - 10 | 1076 | 79 | ||
| U81045.1 | - | 9 - 10 | 945 | 79 | ||
More Information
Set Membership: |
|

Akr1b8
Genomic Map