Assay Details
| Gene Symbol: LOC100760212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291946.1 | NP_001278875.1 | 13 - 14 | 1259 | 66 | |
| XM_003507038.1 | XP_003507086.1 | 13 - 14 | 1271 | 66 | ||
More Information
Set Membership: |
|

LOC100760212
Genomic Map