Assay Details
| Gene Symbol: LOC100760212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291946.1 | NP_001278875.1 | 27 - 28 | 3155 | 63 | |
| XM_003507038.1 | XP_003507086.1 | 27 - 28 | 3167 | 63 | ||
More Information
Set Membership: |
|

LOC100760212
Genomic Map