Assay Details
| Gene Symbol: Abcb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243989.1 | NP_001230918.1 | 23 - 24 | 3090 | 89 | |
| XM_007646774.1 | XP_007644964.1 | 24 - 25 | 3254 | 89 | ||
| XM_007646775.1 | XP_007644965.1 | 22 - 23 | 3029 | 89 | ||
| GenBank mRNA | M17896.1 | - | 10 - 11 | 1219 | 89 | |
| M60041.1 | - | 23 - 24 | 3090 | 89 | ||
More Information
Set Membership: |
|

Abcb1b
Genomic Map