Assay Details
| Gene Symbol: Ercc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244032.1 | NP_001230961.1 | 10 - 11 | 2026 | 57 | |
| GenBank mRNA | AB017635.1 | - | 10 - 11 | 2054 | 57 | |
| EU492902.1 | - | 10 - 11 | 2012 | 57 | ||
More Information
Set Membership: |
|

Ercc4
Genomic Map