Assay Details
| Gene Symbol: Mgat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244074.1 | NP_001231003.1 | 1 | 1521 | 96 | |
| GenBank mRNA | AY598727.1 | - | 1 | 1521 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mgat3| Gene Symbol: Mgat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244074.1 | NP_001231003.1 | 1 | 1521 | 96 | |
| GenBank mRNA | AY598727.1 | - | 1 | 1521 | 96 | |
Set Membership: |
|