Assay Details
| Gene Symbol: B4galt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246691.1 | NP_001233620.1 | 1088 | 87 | ||
| GenBank mRNA | AF318896.1 | - | 1088 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

B4galt1| Gene Symbol: B4galt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246691.1 | NP_001233620.1 | 1088 | 87 | ||
| GenBank mRNA | AF318896.1 | - | 1088 | 87 | ||
Set Membership: |
|