Assay Details
| Gene Symbol: G6pd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246727.1 | NP_001233656.1 | 9 - 10 | 1185 | 62 | |
| GenBank mRNA | AF044676.1 | - | 9 - 10 | 1185 | 62 | |
| JF935143.1 | - | 8 - 9 | 1050 | 62 | ||
| JI880984.1 | - | 4 - 5 | 565 | 62 | ||
| JP058834.1 | - | 4 - 5 | 565 | 62 | ||
More Information
Set Membership: |
|

G6pd
Genomic Map