Assay Details
| Gene Symbol: Cdc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246772.1 | NP_001233701.1 | 11 - 12 | 1527 | 64 | |
| XM_007649581.1 | XP_007647771.1 | 11 - 12 | 1532 | 64 | ||
| XM_007649582.1 | XP_007647772.1 | 10 - 11 | 1436 | 64 | ||
| XM_007649583.1 | XP_007647773.1 | 11 - 12 | 1367 | 64 | ||
| GenBank mRNA | AF281019.1 | - | 11 - 12 | 1527 | 64 | |
| JI894278.1 | - | 2 - 3 | 231 | 64 | ||
| JP040830.1 | - | 11 - 12 | 1502 | 64 | ||
More Information
Set Membership: |
|

Cdc7
Genomic Map