Assay Details
| Gene Symbol: Tubb4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244098.1 | NP_001231027.1 | 1 - 2 | 217 | 80 | |
| GenBank mRNA | JI869795.1 | - | 1 - 2 | 239 | 80 | |
| JP055125.1 | - | 1 - 2 | 239 | 80 | ||
| X60785.1 | - | 1 - 2 | 217 | 80 | ||
More Information
Set Membership: |
|

Tubb4b
Genomic Map