Assay Details
| Gene Symbol: Adk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244850.1 | NP_001231779.1 | 5 - 6 | 473 | 56 | |
| GenBank mRNA | JI869933.1 | - | 4 - 5 | 376 | 56 | |
| JP047183.1 | - | 4 - 5 | 375 | 56 | ||
| U26588.1 | - | 5 - 6 | 473 | 56 | ||
More Information
Set Membership: |
|

Adk
Genomic Map