Assay Details
| Gene Symbol: Rcan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246731.1 | NP_001233660.1 | 1 - 2 | 154 | 65 | |
| GenBank mRNA | JI877057.1 | - | 1 - 2 | 270 | 65 | |
| JP065307.1 | - | 1 - 2 | 269 | 65 | ||
| U60263.1 | - | 1 - 2 | 154 | 65 | ||
More Information
Set Membership: |
|

Rcan1
Genomic Map