Assay Details
| Gene Symbol: Dbf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246771.1 | NP_001233700.1 | 8 - 9 | 1001 | 63 | |
| GenBank mRNA | AF292400.1 | - | 8 - 9 | 1001 | 63 | |
| JI878940.1 | - | 8 - 9 | 888 | 63 | ||
| JP063296.1 | - | 8 - 9 | 888 | 63 | ||
More Information
Set Membership: |
|

Dbf4
Genomic Map