Assay Details
| Gene Symbol: Rbm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246799.1 | NP_001233728.1 | 4 - 5 | 413 | 64 | |
| XM_007645600.1 | XP_007643790.1 | 5 - 6 | 531 | 64 | ||
| GenBank mRNA | EU556497.1 | - | 4 - 5 | 413 | 64 | |
| JI874849.1 | - | 6 - 7 | 939 | 64 | ||
More Information
Set Membership: |
|

Rbm3
Genomic Map