Assay Details
| Gene Symbol: Gstp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246744.1 | NP_001233673.1 | 5 - 6 | 444 | 97 | |
| GenBank mRNA | JI886703.1 | - | 6 - 7 | 543 | 97 | |
| JP038870.1 | - | 6 - 7 | 543 | 97 | ||
| L40381.1 | - | 5 - 6 | 444 | 97 | ||
More Information
Set Membership: |
|

Gstp1
Genomic Map