Assay Details
| Gene Symbol: Rnaseh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285919.1 | NP_001272848.1 | 3 - 4 | 408 | 64 | |
| XM_007650354.1 | XP_007648544.1 | 3 - 4 | 430 | 64 | ||
| XM_007650355.1 | XP_007648545.1 | 3 - 4 | 311 | 64 | ||
| XM_007650356.1 | XP_007648546.1 | 2 - 3 | 252 | 64 | ||
| GenBank mRNA | AB604326.1 | - | 3 - 4 | 408 | 64 | |
| JI871966.1 | - | 2 - 3 | 239 | 64 | ||
| JP058577.1 | - | 2 - 3 | 239 | 64 | ||
More Information
Set Membership: |
|

Rnaseh1
Genomic Map