Assay Details
| Gene Symbol: LOC100689011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243997.1 | NP_001230926.1 | 6 - 7 | 1047 | 60 | |
| XM_007646346.1 | XP_007644536.1 | 6 - 7 | 1486 | 60 | ||
| XM_007646348.1 | XP_007644538.1 | 6 - 7 | 1471 | 60 | ||
| XM_007646350.1 | XP_007644540.1 | 6 - 7 | 1134 | 60 | ||
| XM_007646351.1 | XP_007644541.1 | 6 - 7 | 1119 | 60 | ||
| GenBank mRNA | AY064388.1 | - | 6 - 7 | 1047 | 60 | |
More Information
Set Membership: |
|

LOC100689011
Genomic Map