Assay Details
| Gene Symbol: Dnase1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244118.1 | NP_001231047.1 | 4 - 5 | 588 | 71 | |
| XM_007652793.1 | XP_007650983.1 | 3 - 4 | 654 | 71 | ||
| GenBank mRNA | DQ116786.1 | - | 4 - 5 | 588 | 71 | |
More Information
Set Membership: |
|

Dnase1l1
Genomic Map